Learn More
Cayman Chemical TenofovIr-d6 500ug
SDP
Supplier:  Cayman Chemical 21071500 UG
Encompass_Preferred
An internal standard for the quantification of tenofovir by GC- or LC-MS
Catalog No. 50-298-5566