Learn More
Medchemexpress LLC Tenofovir disoproxil | 201341-05-1 | >99.5% | 519.44 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY13782A100MG
Encompass_Preferred
Tenofovir disoproxil (Bis(POC)-PMPA) is a nucleotide reverse transcriptase inhibitor. It is used to treat HIV and chronic Hepatitis B infections. This compound is for research use only.
  • Nucleotide reverse transcriptase inhibitor
  • Used to treat HIV
  • Used to treat chronic Hepatitis B
  • For research use only
Catalog No. 50-003-50981