Learn More
Medchemexpress LLC Tenofovir Disoproxil | 201341-05-1 | 99.5% | 519.44 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY13782100MG
Encompass_Preferred
Tenofovir Disoproxil is a nucleotide reverse transcriptase inhibitor. It is used to treat HIV and chronic Hepatitis B, and is intended for research use only.
  • Nucleotide reverse transcriptase inhibitor
  • Used to treat HIV
  • Used to treat chronic Hepatitis B
  • Solid appearance
  • White to off-white color
  • Purity 99.5%
  • For research use only
Catalog No. 50-003-49643