Learn More
Medchemexpress LLC Tenofovir Disoproxil | 201341-05-1 | 99.5% | 519.44 | 50 MG

Supplier: Medchemexpress LLC HY13782A50MG
Tenofovir Disoproxil is a nucleotide reverse transcriptase inhibitor primarily used in the treatment of HIV and chronic Hepatitis B. This product is intended for research use only and is not sold to patients, providing a valuable tool for scientific studies.
- Nucleotide reverse transcriptase inhibitor
- Aids in the treatment of HIV and chronic Hepatitis B
- High purity of 99.5%
- Provides in vitro and in vivo solubility data
- Stable in powder form for up to 3 years at -20°C
- Stable in solvent for up to 2 years at -80°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.