Learn More
TARGETMOL CHEMICALS INC Tenofovir Disoproxil 500MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T7111500MG
Encompass_Preferred
Also available in 1 g, 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Tenofovir Disoproxil (GS 4331) is a nucleotide reverse transcriptase inhibitor to treat HIV and chronic Hepatitis B. Purity 99.17%
Catalog No. 50-287-8137