Learn More
Medchemexpress LLC Tenofovir Disoproxil (fumarate) | 202138-50-9 | 99.7% | 635.51 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY13782R100MG
Encompass_Preferred
Tenofovir Disoproxil fumarate is a nucleotide reverse transcriptase inhibitor employed in the treatment of HIV and chronic Hepatitis B. This compound demonstrates significant antiretroviral and anti-HBV activity, making it a crucial agent in managing these viral infections.
  • Potent nucleotide reverse transcriptase inhibitor
  • Effective against HIV and chronic Hepatitis B
  • High purity of 99.7%
  • Targets HIV and HBV through anti-infection pathways
  • Demonstrates cytotoxic and anti-viral effects in cellular models
  • Proven to reduce HIV transmission in in vivo studies
  • Induces dose-dependent decline in viremia in chronic HBV models
Catalog No. 50-003-51950