Learn More
Medchemexpress LLC Tenofovir disoproxil (fumarate) | 202138-50-9 | MFCD08141829 | >98.0% | C23H34N5O14P | 10MG
SDP
Supplier:  Medchemexpress LLC HY13782R10MG
Encompass_Preferred
Tenofovir disoproxil fumarate (CAS 202138-50-9) is the fumarate salt of the tenofovir prodrug and an acyclic nucleotide analog reverse transcriptase inhibitor used as a research chemical for antiviral and reverse transcriptase studies. MedChemExpress supplies this research-grade compound with supporting safety and specification documents for laboratory use.
  • Acts as a prodrug of tenofovir, a nucleotide reverse transcriptase inhibitor.
  • Intended for antiviral and reverse transcriptase research applications.
  • Provided as a research-grade solid with supporting safety documentation.
  • High purity suitable for analytical and biological assays.
  • Compatible with common laboratory solvents for dissolution and formulation.
Catalog No. 50-002-61769