Learn More
Medchemexpress LLC Tenofovir disoproxil (fumarate) | 202138-50-9 | MFCD08141829 | 99.7% | 635.51 | C23H34N5O14P | 5 MG
SDP
Supplier:  Medchemexpress LLC HY13782R5MG
Encompass_Preferred
Tenofovir disoproxil fumarate (Standard) is the fumarate salt of tenofovir disoproxil provided as a high-purity analytical standard for research and quality control applications. It is intended for method development, assay validation, and use as a reference material in studies of tenofovir prodrugs and antiviral activity.
  • High purity suitable for HPLC and LC-MS analyses.
  • Supplied in small solid packs for analytical workflows.
  • Stable solid form for reliable reference measurements.
  • Includes a certificate of analysis and safety data sheet for traceability.
  • Supports method validation and quality control of tenofovir-related assays.
Catalog No. 50-002-22017