Learn More
Selleck Chemical LLC Tenofovir Disoproxil Fumarate S1400-1g
SDP
Supplier:  Selleck Chemical LLC S14001G
Encompass_Preferred
Tenofovir Disoproxil Fumarate belongs to a class of antiretroviral drugs it inhibits the activity of HIV reverse transcriptase by competing with the natural substrate deoxyadenosine 5 -triphosphate and after incorporation into DNA by DNA chain termination
Catalog No. 50-311-1408