Learn More
Medchemexpress LLC Tenofovir (S-enantiomer) | 147127-19-3 | MFCD21604708 | 98.3% | 287.21 g/mol | C9H14N5O4P | 1 ML

Supplier: Medchemexpress LLC HYW07493010MM/1ML
(S)-Tenofovir is the S-enantiomer of tenofovir, a nucleotide reverse transcriptase inhibitor used in research on HIV and chronic hepatitis B. The product is supplied for research use and is available as a 10 mM solution in DMSO (1 mL) or as a solid in multiple mass sizes.
- S-enantiomer of tenofovir used in mechanistic and antiviral research
- Nucleotide reverse transcriptase inhibitor active against HIV and HBV
- Available as 10 mM solution in DMSO (1 mL) and solid formats (10 mg-500 mg)
- Chemical formula C9H14N5O4P and molecular weight 287.21 g/mol
- Reported purity approximately 98.3%
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.