Learn More
Apexbio Technology LLC Tenovin-1, 25mg. Cas: 380315-80-0 MFCD: MFCD02914760. SIRT2 inhibitor, activates p53
SDP
Supplier:  Apexbio Technology LLC A420325
Encompass_Preferred
Tenovin-1, a small molecule discovered by a cell-based screen, is a bio-active activator of p53 that elevates the levels of p53 protein, p53-downstream target p21CIP/WAF1 protein and mRNA and protects p53 from mdm2-mediated degradation with little impact on p53 synthesis. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4904