Learn More
Medchemexpress LLC HY-15510 5mg Medchemexpress, Tenovin-6 CAS:1011557-82-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY155105MG
Encompass_Preferred
Medchemexpress, HY-15510 5mg Tenovin-6 CAS:1011557-82-6 Tenovin-6, an analog of Tenovin-1 (HY-13423), is an activator of p53 transcriptional activity. Tenovin-6 inhibits the protein deacetylase activities of purified human SIRT1, SIRT2, and SIRT3 with IC50s of 21 μM, 10 μM, and 67 μM, respectively. Tenovin-6 also inhibits dihydroorotate dehydrogenase (DHODH). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1728