Learn More
Medchemexpress LLC Tenovin-6 hydrochloride | 1011301-29-3 | 98.9% | 491.09 g·mol^-1 | C25H35ClN4O2S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15510B10MG
Encompass_Preferred
Tenovin-6 hydrochloride is a research-grade small-molecule p53 activator and sirtuin deacetylase inhibitor used in biochemical and cellular studies to modulate p53 signaling and protein acetylation. Supplied as a hydrochloride salt to aid solubility, it is intended for in vitro research applications where modulation of SIRT1/2/3 activity is required.
  • Activates p53 transcriptional activity in cellular assays.
  • Inhibits SIRT1, SIRT2, and SIRT3 deacetylase activity.
  • Hydrochloride salt form improves aqueous solubility for in vitro use.
  • Available in small research quantities for laboratory studies.
  • Suitable for biochemical and cell-based assay applications.
  • Provided with a product datasheet for handling and storage guidance.
Catalog No. 50-002-86835