Learn More
Medchemexpress LLC Tentoxin-d3 | 1426437-58-2 | 417.52 | C22H27D3N4O4 | 250 UG
SDP
Supplier:  Medchemexpress LLC HYW653894250UG
Encompass_Preferred
Tentoxin-d3 is the deuterium-labeled analogue of tentoxin, provided as a small-quantity research standard for analytical applications such as mass spectrometry and metabolic tracing. Molecular formula C22H27D3N4O4; molecular weight 417.52; CAS 1426437-58-2.
  • Deuterium-labeled (d3) analogue for use as an internal standard in mass spectrometry
  • Suitable for metabolic tracing and analytical method development
  • Provided in a low microgram quantity for sensitive assays
  • Molecular weight 417.52; molecular formula C22H27D3N4O4
  • Store powder at -20°C for long-term stability
Catalog No. 50-002-84967