Learn More
Selleck Chemical LLC Tenuifolin S9087-1MG
SDP
Supplier:  Selleck Chemical LLC S90871MG
Encompass_Preferred
Tenuifolin a secondary saponin isolated from hydrolysates of polygalasaponins inhibits beta-amyloid synthesis in vitro and has nootropic activity in vivo via acetylcholine esterase inhibition and increases norepinephrine and dopamine production
Catalog No. 50-000-24126