Learn More
Medchemexpress LLC Tenuifoliside C | 139726-37-7 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN25865MG
Encompass_Preferred
Tenuifoliside C, isolated from *Polygala tenuifolia Willd*, is known for its significant inhibitory effect on chlorzoxazone 6-hydroxylation catalyzed by CYP2E1. This compound has also demonstrated various biological activities, including cytotoxicity against several human cancer cell lines and anti-inflammatory effects by inhibiting LPS-induced nitric oxide production.
  • Significantly inhibits chlorzoxazone 6-hydroxylation catalyzed by CYP2E1.
  • Exhibits cytotoxicity against human A549, HCT15, SK-MEL-2, and SK-OV-3 cells.
  • Demonstrates anti-inflammatory properties by inhibiting nitric oxide production.
Catalog No. 50-004-18465