Learn More
Medchemexpress LLC Tepotinib | 1100598-32-0 | 99.9% | 492.57 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1472150MG
Encompass_Preferred
Tepotinib is an orally active, highly selective, reversible, and ATP-competitive inhibitor of c-Met with an IC50 of 3 nM. It demonstrates over 200-fold selectivity for c-Met compared to IRAK4, TrkA, Axl, IRAK1, and Mer. This compound inhibits c-Met phosphorylation and induces autophagy, exhibiting antitumor effects.
  • Orally active
  • Highly selective c-Met inhibitor
  • Reversible and ATP-competitive mechanism
  • Inhibits c-Met phosphorylation
  • Induces autophagy
  • Demonstrates antitumor effects
Catalog No. 50-003-61982