Learn More
Selleck Chemical LLC TEPP-46 (ML265) 5mg 1221186-53-3 CID-44246499, NCGC00186528
SDP
Supplier:  Selleck Chemical LLC S73025MG
Encompass_Preferred
TEPP-46 (ML265, CID-44246499, NCGC00186528) is a potent activator of PKM2 in both biochemical (AC50 = 92 nM) and cell-based assays with high selectivity over PKM1, PKR and PKL. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8291