Learn More
Apexbio Technology LLC Terbinafine, 100mg. Cas: 91161-71-6 MFCD: MFCD00242672. Allylamine antifungal agent
SDP
Supplier:  Apexbio Technology LLC A8533100
Encompass_Preferred
Terbinafine is an allylamine antifungal agent which has fungicidal activity against a wide variety of dermatophytes, moulds and certain dimorphic fungi, and fungistatic activity against Candida albicans. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5157