Learn More
Medchemexpress LLC Terbinafine-d3 hydrochloride | 1310012-15-7 | 99.87% | 330.91 | 1 MG
Supplier: Medchemexpress LLC HY17395S1MG
Terbinafine-d3 hydrochloride is the deuterium-labeled Terbinafine hydrochloride. It is an antifungal medication utilized to treat fungal infections. It acts as a potent non-competitive inhibitor of squalene epoxidase from Candida with a Kᵢ of 30 nM. It also exhibits antibacterial activity against certain gram-positive and gram-negative bacteria.
- Deuterium labeled.
- Antifungal medication.
- Potent non-competitive inhibitor of squalene epoxidase.
- Exhibits antibacterial activity against certain gram-positive and gram-negative bacteria.
- Stable heavy isotopes of hydrogen, carbon, and other elements incorporated into drug molecules can act as tracers for quantitation during drug development.
- Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
- Can be used as a tracer.
- Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.