Learn More
Enzo Life Sciences Terbinafine HCl (100mg). CAS: 78628-80-5
SDP
Supplier:  Enzo Life Sciences BMLEI3180100
Encompass_Preferred
Inhibits ergosterol biosynthesis at the level of squalene epoxidase. Potent non-competitive inhibitor of squalene epoxidase from Candida (Ki=30 nM), inhibiting mammalian enzymes at higher concentrations (4-77 µM). It inhibits HUVEC proliferation and angiogenesis and displays interesting anti-inflammatory and free radical scavenging activities. Clinically useful antifungal agent. Alternative name: trans-N-(6,6-Dimethyl-2-hepten-4-ynyl)-N-methyl-1-naphthylmethylamine . hydrochloride. Purity: ≥98% (UHPLC). Solubility: Soluble in ethanol (45 mg/ml), DMSO (30 mg/ml) or water (3 mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0707