Learn More
Selleck Chemical LLC Terbinafine HCl-50mg
SDP
Supplier:  Selleck Chemical LLC S255750MG
Encompass_Preferred
Terbinafine HCl (KWD 201,KWD 2019,TDT 067 hydrochloride) inhibits ergosterol synthesis by inhibiting squalene epoxidase, used as an antifungal drug.
Catalog No. 50-238-9991