Learn More
Strem, An Ascensus Company CAS# 7440-27-9. 5g. Terbium powder (99.9% REO). MFCD00011256
SDP
Supplier:  Strem, An Ascensus Company 9365155G
Encompass
CAS# 7440-27-9. 5g. Terbium powder (99.9% REO). MFCD00011256. Molecular Weight: 158.92. Molecular Formula: Tb. Color/form: -40 mesh Strem# 93-6515. http://www.strem.com/catalog/v/93-6515/
Catalog No. 50-901-16097