Learn More
Selleck Chemical LLC Terbutaline E4849-25MG
SDP
Supplier:  Selleck Chemical LLC E484925MG
Encompass_Preferred
Terbutaline(Brican Brethine) an active metabolite of bambuterol is an orally active agonist of 2-adrenergic receptor that increases MKP-1 expression Terbutaline is used in the treatment of obstructive lung diseases as a bronchodilator remedy It also exhibits anti-inflammatory effects
Catalog No. 50-000-42608