Learn More
Apexbio Technology LLC Terbutaline Sulfate 23031-32-5 10mM (in 1mL H2O)
SDP
Supplier:  Apexbio Technology LLC B13285.1
Encompass_Preferred
Terbutaline Sulfate is a selective agonist targeting 2-adrenergic receptors known to activate downstream intracellular signal transduction cascades via adenylate cyclase stimulation Upon receptor binding it increases intracellular cyclic adenosine monophosphate (cAMP) levels subsequently leading to smooth muscle relaxation in bronchial tissues and vasculature Due to this specific 2-adrenoceptor activity Terbutaline Sulfate frequently serves as a research tool in studies investigating bronchial responsiveness airway hyperreactivity asthma pathophysiology and smooth muscle regulatory mechanisms Researchers utilize this molecule to elucidate receptor-mediated molecular pathways assess receptor binding kinetics and perform functional assays evaluating -adrenergic receptor signaling and cellular responses
Catalog No. 50-000-62305