Learn More
Medchemexpress LLC Terbutaline (sulfate) | 23031-32-5 | 100.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB080210MM/1ML
Encompass_Preferred
Terbutaline sulfate is an orally active β2-adrenergic receptor agonist and an active metabolite of bambuterol. This compound is primarily used in asthma symptom research.
  • Orally active β2-adrenergic receptor agonist
  • Active metabolite of bambuterol
  • Used in asthma symptom research
  • Enhances MKP-1 expression in activated mouse macrophages (in vitro studies)
  • Improves allodynia in ob/ob mice (in vivo studies)
Catalog No. 50-004-08115