Learn More
Medchemexpress LLC Terbuthylazine-d5 | 222986-60-9 | 99.1% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB1847S11MG
Encompass_Preferred
Terbuthylazine-d5 is a deuterium labeled version of Terbuthylazine, which acts as an inhibitor of acetolactate synthase (ALS) and is a selective herbicide. Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-10800