Learn More
Medchemexpress LLC HY-W010098 5g Medchemexpress, Terephthalic acid CAS:100-21-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW0100985G
Encompass_Preferred
Medchemexpress, HY-W010098 5g Terephthalic acid CAS:100-21-0 Terephthalic acid is one isomer of the three phthalic, a precursor to the polyester PET, used to make clothing and plastic bottles. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4531