Learn More
Medchemexpress LLC Terephthalic acid-d4 | 60088-54-2 | 98.0% | 170.16 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW010098S50MG
Encompass_Preferred
Terephthalic acid-d4 is the deuterium-labeled form of Terephthalic acid, an isomer and a precursor to polyester PET, a material used in clothing and plastics. It can also be utilized to construct skin tumor models.
  • Can be used as a tracer.
  • Can serve as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Deuteration, the incorporation of stable heavy isotopes like deuterium into drug molecules, has shown potential in influencing the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-51557