Learn More
Sigma Aldrich Fine Chemicals Biosciences TERGITOL™ solution, MFCD01779855, 100mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences NP40S100ML
Encompass
Type NP-40, 70% in H2O, Tergitol solution is a surface-active agent that acts as a non-ionic surfactant.
Catalog No. NC1618930