Learn More
Medchemexpress LLC Terlipressin diacetate | 1884420-36-3 | 99.8% | 1347.48 | C56H82N16O19S2 | 100MG
SDP
Supplier:  Medchemexpress LLC HY12554A100MG
Encompass_Preferred
Terlipressin diacetate is a vasopressin analogue and a selective vasopressin V1 receptor agonist used in research to reduce splanchnic blood flow and portal pressure. It is applied in studies of acute variceal bleeding, hepatorenal syndrome, and norepinephrine-resistant septic shock, and shows anti-inflammatory and anti-oxidative effects. Supplied as a purified peptide with recommended frozen storage to preserve stability.
  • High purity suitable for research applications.
  • Selective vasopressin V1 receptor agonist activity.
  • Relevant for models of portal hypertension and variceal bleeding.
  • Exhibits anti-inflammatory and anti-oxidative properties.
  • Storage recommendations minimize degradation and extend shelf life.
  • Available in multiple pack sizes for flexible experimental needs.
Catalog No. 50-002-35999