Learn More
Medchemexpress LLC Terlipressin diacetate | 1884420-36-3 | 99.8% | 1347.48 | C56H82N16O19S2 | 25MG

Supplier: Medchemexpress LLC HY12554A25MG
Terlipressin diacetate is a synthetic vasopressin V1 receptor agonist provided as a high-purity solid peptide for laboratory research. It is typically used to investigate vasoconstriction, portal pressure modulation, and related pharmacological effects. The product includes storage and stability guidance for both powder and solution forms.
- High purity (99.76%) suitable for research use.
- Molecular weight 1347.48 and defined molecular formula.
- Available in multiple pack sizes from 5 mg to 500 mg.
- Storage conditions and stability provided for powder and in-solution forms.
- Supplied as a solid peptide salt appropriate for pharmacology studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.