Learn More
Medchemexpress LLC Terlipressin diacetate | 1884420-36-3 | 99.8% | 1347.48 | C56H82N16O19S2 | 50MG
SDP
Supplier:  Medchemexpress LLC HY12554A50MG
Encompass_Preferred
Terlipressin diacetate is a synthetic vasopressin analogue and selective vasopressin V1 receptor agonist used in research to reduce splanchnic blood flow and lower portal pressure. It is investigated for control of acute variceal bleeding, hepatorenal syndrome, and norepinephrine-resistant septic shock. Supplied as a white to off-white solid, with molecular weight 1347.48, formula C56H82N16O19S2, and reported purity ~99.8%.
  • Selective vasopressin V1 receptor agonist.
  • Reduces splanchnic blood flow and portal pressure.
  • Suitable for research on variceal bleeding and hepatorenal syndrome.
  • High reported purity for analytical and pharmacological studies.
  • Solid form facilitates accurate dosing and storage.
  • Stable under recommended low-temperature storage conditions.
Catalog No. 50-002-39298