Learn More
Cayman Chemical Terpendole I 1mg
SDP
Supplier:  Cayman Chemical 294431 MG
Encompass_Preferred
A fungal metabolite; a weak ACAT inhibitor (IC50 = 145 µM); active against the bacteria B. cereus and B. subtilis (MICs = 100 UG/ml for both) but not S. aureus, P. aeruginosa, or K. pneumoniae (MICs = >200 UG/ml for all) or the fungus C. albicans (MIC = >200 UG/ml); cytotoxic to HeLa cells (IC50 = 52.6 µM)
Catalog No. 50-298-8274