Learn More
Cayman Chemical TerpIn-4-ol 500mg
SDP
Supplier:  Cayman Chemical 38964500 MG
Encompass_Preferred
A monoterpene alcohol with arrhythmogenic and anticonvulsant activities; induces extrasystoles in isolated rat left atria at 100 µM; induces supraventricular tachycardia in rats at 10 mg/kg; increases the latency to seizure and decreases the number of seizures in a rat model of PTZ-induced epilepsy
Catalog No. 50-291-6040