Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Terpin hydrate | 2451-01-6 | 98.0% | 500 MG
SDP

Supplier:  Medchemexpress LLC HYB1063500MG

Encompass_Preferred

Terpin hydrate is an expectorant agent used in the research of mucus associated with acute and chronic bronchitis. It has been shown to ameliorate lung damage induced by paraquat dichloride.

  • Used in research for acute and chronic bronchitis.
  • Shown to reduce paraquat dichloride-induced lung damage.
  • Solid form, white to off-white in color.
  • Purity of 98.0%.
  • Molecular weight is 190.28.
  • Store powder at -20°C for 3 years or 4°C for 2 years.
  • Store in solvent at -80°C for 2 years or -20°C for 1 year.

Catalog No. 50-004-12759


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.