Learn More
Cayman Chemical Terpn-4-ol 5g
SDP
Supplier:  Cayman Chemical 389645 G
Encompass_Preferred
A monoterpene alcohol with arrhythmogenic and anticonvulsant activities; induces extrasystoles in isolated rat left atria at 100 µM; induces supraventricular tachycardia in rats at 10 mg/kg; increases the latency to seizure and decreases the number of seizures in a rat model of PTZ-induced epilepsy
Catalog No. 50-291-8452