Learn More
Growcells TERRIFIC BROTH 1L CS12
Supplier:  Growcells MBLE327012
Encompass_Preferred
Ready to use sterile bacterial culture media, sterile filtered and steam sterilized in TriForest polycarbonate bottles. Recipe/L: Yeast Extract 24g, Tryptone 12g, K2HPO4 9.4g, KH2PO4 2.2g pH 7.2
Catalog No. 50-103-4792