Learn More
Cayman Chemical TerrItrem A 2.5mg
SDP
Supplier:  Cayman Chemical 299412.5 MG
Encompass_Preferred
A mycotoxin originally isolated from A. terreus; irreversibly inhibits AChE (IC50 = 24 nM for the electric eel enzyme); induces tremors in mice with a median tremulous dose of 0.31 mg/kg; LD50 = 17.6 mg/kg
Catalog No. 50-299-2335