Learn More
Abcam Territrem B, AChE inhibitor, 1MG
Supplier:  Abcam AB1443701MG
Encompass_Preferred
MW 526.6 Da, Purity >95%. Potent and selective irreversible AChE inhibitor (IC₅₀ = 8 nM). Binds to the P-site and A-site of AChE. Potentiates ACh-induced neuronal current. Shows antifungal and cholinergic effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5837