Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (tert-butoxycarbonyl)-D-histidine | 50654-94-9 | MFCD00037851 | 99.3% | 255.27 g/mol | C11H17N3O4 | 10 G
SDP

Supplier:  Medchemexpress LLC HYW05224610G

Encompass_Preferred

(tert-Butoxycarbonyl)-D-histidine is a Boc-protected D-histidine derivative supplied as a white to off-white solid for use as a research reagent and peptide synthesis intermediate.

  • Cas number: 50654-94-9.
  • Molecular formula: C11H17N3O4.
  • Molecular weight: 255.27 g/mol.
  • Purity: 99.3%.
  • Appearance: solid, white to off-white.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solution -80°C (6 months) or -20°C (1 month).
  • Solubility: DMSO 8.33 mg/mL (32.63 mM); may require sonication.
  • Available sizes: 5 g, 10 g, 50 g.
  • Intended use: research reagent; for research use only.

Catalog No. 50-002-09979


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.