Learn More
Medchemexpress LLC Tert-butoxycarbonyl glycylglycylglycine | 28320-73-2 | MFCD00190687 | 289.29 g/mol | C11H19N3O6 | 10 G

Supplier: Medchemexpress LLC HYW04073410G
(tert-Butoxycarbonyl)glycylglycylglycine is a Boc-protected triglycine derivative used as a research reagent in chemical and biochemical studies.
- Used as a Boc-protected peptide building block for synthesis and modification.
- Molecular formula C11H19N3O6.
- Molecular weight 289.29 g/mol.
- Solid powder form suitable for storage and handling in research laboratories.
- Useful in peptide chemistry and other chemical studies requiring protected glycine oligomers.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.