Learn More
Medchemexpress LLC Tert-butoxycarbonyl glycylglycylglycine | 28320-73-2 | MFCD00190687 | ≥97.0% | 289.29 g/mol | C11H19N3O6 | 100 G

Supplier: Medchemexpress LLC HYW040734100G
(tert-Butoxycarbonyl)glycylglycylglycine is a Boc-protected triglycine peptide used as a research reagent and an intermediate in peptide synthesis and biochemical studies. It is supplied as a white to off-white solid with high purity suitable for laboratory use.
- Tert-butoxycarbonyl-protected triglycine
- Used as a peptide synthesis intermediate and research reagent
- CAS number 28320-73-2
- Molecular formula C11H19N3O6, molecular weight 289.29 g/mol
- Appearance: white to off-white solid
- Purity: ≥97.0%
- Available in 100 G packaging
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.