Learn More
Medchemexpress LLC (tert-Butoxycarbonyl)glycylglycylglycine | 28320-73-2 | MFCD00190687 | ≥97.0% | 289.29 g/mol | C11H19N3O6 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW040734
Encompass_Preferred
(tert-Butoxycarbonyl)glycylglycylglycine is a Boc-protected triglycine peptide fragment used as a peptide building block and reagent in organic and peptide synthesis. It provides a protected N-terminal amine that can be deprotected under mild acidic conditions to yield the free amine, and it is supplied with batch analysis confirming high purity.
  • Boc-protected N-terminal amine for selective deprotection under mild acidic conditions.
  • Triglycine fragment suitable as a peptide building block and linker component.
  • High analytical purity as confirmed by batch certificate of analysis.
  • Compatible with standard peptide coupling and synthesis protocols.
  • Available in laboratory pack sizes for synthetic applications.
Catalog No. 50-002-08881