Learn More
Medchemexpress LLC (tert-butoxycarbonyl)-L-leucylglycine | 32991-17-6 | MFCD00076954 | 97.0% | 288.34 g·mol⁻¹ | C13H24N2O5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW039759100MG
Encompass_Preferred
(tert-butoxycarbonyl)-L-leucylglycine is an N-protected dipeptide used as a building block in peptide synthesis and as a reference for analytical method development. It is provided as a powder with molecular formula C13H24N2O5 and molecular weight 288.34 g·mol⁻¹. Consult the certificate of analysis for batch-specific purity and handling instructions.
  • N-protected dipeptide suitable for solid-phase and solution-phase peptide synthesis.
  • Serves as a building block for peptide elongation and derivatization.
  • Supplied as a stable powder for convenient weighing and storage.
  • Compatible with standard peptide coupling reagents and protocols.
  • Useful as an analytical reference in method development and quality control.
  • Storage at low temperature extends shelf life; consult the COA for purity and storage details.
Catalog No. 50-002-99764