Learn More
Medchemexpress LLC Tert-butoxycarbonyl-L-valyl-L-alanine | 70396-18-8 | MFCD22381612 | 98.0% | 288.34 | C13H24N2O5 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0387035G
Encompass_Preferred
tert-Butoxycarbonyl-L-valyl-L-alanine is a Boc-protected dipeptide derivative of L-valine and L-alanine supplied for research use. It is described as an alanine derivative for biochemical and peptide research, with recorded identifiers for compound registration and quality control (CAS 70396-18-8, C13H24N2O5, MW 288.34).
  • Suitable for peptide synthesis and biochemical research applications.
  • Available in multiple sizes for experimental flexibility.
  • High purity confirmed by batch Certificate of Analysis (example: 98.0% by NMR).
  • Includes CAS number, molecular formula, and molecular weight for compound tracking.
  • Provided with batch-specific analytical documentation for quality assurance.
Catalog No. 50-003-01169