Learn More
Medchemexpress LLC Tert-butoxycarbonyl-L-valyl-L-valine | 69209-73-0 | 97.0% | 316.39 g/mol | C15H28N2O5 | 10 G

Supplier: Medchemexpress LLC HYW04883110G
(tert-Butoxycarbonyl)-L-valyl-L-valine is a Boc-protected dipeptide derived from two valine residues, used as a building block in peptide synthesis and related biochemical research applications.
- Boc-protected L-valyl-L-valine dipeptide for peptide synthesis.
- White to off-white powder; molecular weight 316.39 g/mol.
- Chemical formula C15H28N2O5; CAS number 69209-73-0.
- Purity 97.0%.
- Typical package: 10 G; powder storage -20°C (3 years), 4°C (2 years).
- In solvent storage -80°C (6 months), -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.