Learn More
Medchemexpress LLC (Tert-butoxycarbonyl)-l-valyl-l-valine | 69209-73-0 | MFCD00237584 | 97.0% | 316.39 | C15H28N2O5 | 25 G

Supplier: Medchemexpress LLC 25GHYW048831
(tert-Butoxycarbonyl)-L-valyl-L-valine is a Boc-protected dipeptide used as an amino acid derivative building block for peptide synthesis and related biochemical research. It is supplied as a white to off-white solid suitable for laboratory synthesis applications.
- Used as a Boc-protected dipeptide building block in peptide synthesis.
- CAS number 69209-73-0 for unambiguous identification.
- Molecular weight 316.39 and formula C15H28N2O5.
- Purity 97.0% suitable for research use.
- Available in multiple sizes, including 25 G for typical lab use.
- Appearance: solid, white to off-white.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.