Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (tert-butoxycarbonyl)-L-valylglycine | 45233-75-8 | MFCD00134861 | 97.0% | 274.31 | C12H22N2O5 | 10 G
SDP

Supplier:  Medchemexpress LLC HYW03969510G

Encompass_Preferred

(tert-Butoxycarbonyl)-L-valylglycine is a Boc-protected L-valylglycine derivative supplied as a white to off-white solid for use in peptide synthesis and related biochemical research. It has a molecular weight of 274.31 and is provided with documented purity and storage recommendations for reliable laboratory use.

  • Boc-protected dipeptide suitable as a building block in peptide synthesis.
  • White to off-white solid for easy handling and weighing.
  • Molecular weight 274.31 for accurate stoichiometry.
  • Purity 97.0% to support reproducible experimental results.
  • Provided with datasheet, certificate of analysis, and safety data for compliance.
  • Recommended storage conditions to maintain stability.

Catalog No. 50-002-99760


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.