Learn More
Medchemexpress LLC tert-butoxycarbonyl methionine | 93000-03-4 | MFCD00190811 | 95.0% | 249.33 g/mol | C10H19NO4S | 100 G

Supplier: Medchemexpress LLC HYW052309100G
(tert-Butoxycarbonyl)methionine is the N-tert-butoxycarbonyl (Boc) protected derivative of the amino acid methionine. It is supplied as a white to off-white solid and is used as an N-protected building block in peptide synthesis and other organic synthesis applications where temporary protection of the amino terminus is required.
- Used as an N-protected amino acid building block for peptide synthesis.
- Provides tert-butoxycarbonyl (Boc) protection of the amino terminus.
- Supplied as a white to off-white solid for ease of handling and storage.
- High purity suitable for synthetic applications (listed purity 95.0%).
- Characterized by formula C10H19NO4S and molecular weight 249.33 g/mol.
- Documentation available: certificate of analysis and safety data sheet.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.